missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Trifluoromethylphenyl)anthranilic Acid 98.0+%, TCI America™

Catalog No. T235425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T235425G 25 g
T2354500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T235425G Supplier TCI America Supplier No. T235425G

Chemical Identifiers

CAS 530-78-9
Molecular Formula C14H10F3NO2
Molecular Weight (g/mol) 281.234
MDL Number MFCD00002422
InChI Key LPEPZBJOKDYZAD-UHFFFAOYSA-N
Synonym flufenamic acid, fluphenamic acid, nichisedan, achless, arlef, flufacid, fullsafe, lanceat, paraflu, plostene
PubChem CID 3371
ChEBI CHEBI:42638
IUPAC Name 2-[3-(trifluoromethyl)anilino]benzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)NC2=CC=CC(=C2)C(F)(F)F

Specifications

Melting Point 135°C
Quantity 25 g
UN Number 2811
Formula Weight 281.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(3-Trifluoromethylphenyl)anthranilic Acid
EINECSNumber (3)-1493&(3)-3002&(9)-0907
RTECSNumber CB4375000
TSCA No