accessibility menu, dialog, popup

UserName

CAS RN 530-78-9

IUPAC Name: 2-{[3-(trifluoromethyl)phenyl]amino}benzoic acid
Synonyms: flufenamic acid tecramine fluore-200 aluminum flufenamate fullsafe arlef acid, flufenamic reumajust A fluphenamic acid paraflu
Molecular Weight (g/mol): 281.23
Molecular Formula: C14H10F3NO2
InChi Key: LPEPZBJOKDYZAD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1NC1=CC=CC(=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)