missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Pyridyl)thiourea, 98+%

Catalog No. aal0128403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0128403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0128403 Supplier Thermo Scientific Chemicals Supplier No. L0128403
Only null left

CAS: 30162-37-9 | C6H7N3S | 153.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30162-37-9
Molecular Formula C6H7N3S
Molecular Weight (g/mol) 153.20
MDL Number MFCD00014635
InChI Key CFOJQUGXHMGMOT-UHFFFAOYSA-N
Synonym 3-pyridylthiourea, 1-3-pyridyl-2-thiourea, 1-pyridin-3-yl thiourea, pyridin-3-yl-thiourea, n-3-pyridyl thiourea, n-pyridin-3-ylthiourea, pyridin-3-yl thiourea, 1-pyridin-3-ylthiourea, amino 3-pyridylamino methane-1-thione, 3-pyridinylthiourea
PubChem CID 2760515
IUPAC Name pyridin-3-ylthiourea
SMILES NC(=S)NC1=CC=CN=C1

Specifications

Melting Point 158°C to 162°C
Quantity 1 g
UN Number UN2811
Beilstein 472146
Formula Weight 153.21
Percent Purity ≥98%
Chemical Name or Material N-(3-Pyridyl)thiourea
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 250-075-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only