missing translation for 'onlineSavingsMsg'
Learn More

N-[3,5-Bis(trifluoromethyl)phenyl]thiourea, 98+%

Catalog No. p-7024965
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2024503 1 g
AAB2024506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2024503 Supplier Thermo Scientific Chemicals Supplier No. B2024503
Only null left

CAS: 175277-17-5 | C9H6F6N2S | 288.211 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 175277-17-5
Molecular Formula C9H6F6N2S
Molecular Weight (g/mol) 288.211
MDL Number MFCD00041172
InChI Key CWRWOECVPKDZIC-UHFFFAOYSA-N
Synonym 1-3,5-bis trifluoromethyl phenyl-2-thiourea, 1-3,5-bis trifluoromethyl phenyl thiourea, 3,5-bis trifluoromethyl phenylthiourea, 3,5-bis trifluoromethyl phenyl thiourea, n-3,5-di trifluoromethyl phenyl thiourea, n-3,5-bis trifluoromethyl phenyl thiourea, thiourea,n-3,5-bis trifluoromethyl phenyl, amino 3,5-bis trifluoromethyl phenyl amino methane-1-thione, maybridge1_008805
PubChem CID 2736183
IUPAC Name [3,5-bis(trifluoromethyl)phenyl]thiourea
SMILES C1=C(C=C(C=C1C(F)(F)F)NC(=S)N)C(F)(F)F

Specifications

Melting Point 168°C to 172°C
Quantity 1 g
UN Number UN2811
Formula Weight 288.22
Percent Purity ≥98%
Chemical Name or Material N-[3,5-Bis(trifluoromethyl)phenyl]thiourea
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only