accessibility menu, dialog, popup

UserName

CAS RN 175277-17-5

IUPAC Name: [3,5-bis(trifluoromethyl)phenyl]thiourea
Synonyms: 3,5-bis(trifluoromethyl)phenylthiourea
Molecular Weight (g/mol): 288.21
Molecular Formula: C9H6F6N2S
InChi Key: CWRWOECVPKDZIC-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results