missing translation for 'onlineSavingsMsg'
Learn More

N-[3-[3-(Dimethylamino)acryloyl]phenyl]-N-ethylacetamide 98.0+%, TCI America™

Catalog No. D42595G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42595G 5 g
D42591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42595G Supplier TCI America Supplier No. D42595G

Chemical Identifiers

CAS 96605-66-2
Molecular Formula C15H20N2O2
Molecular Weight (g/mol) 260.337
MDL Number MFCD04117958
InChI Key UXWJJVRASIHSQS-MDZDMXLPSA-N
PubChem CID 11149633
IUPAC Name N-[3-[(E)-3-(dimethylamino)prop-2-enoyl]phenyl]-N-ethylacetamide
SMILES CCN(C1=CC=CC(=C1)C(=O)C=CN(C)C)C(=O)C

Specifications

Melting Point 113°C
Color Yellow
Quantity 5 g
Formula Weight 260.34
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[3-[3-(Dimethylamino)acryloyl]phenyl]-N-ethylacetamide