accessibility menu, dialog, popup

UserName

CH3NONCH3H3COCH3(E/Z)

CAS RN 96605-66-2

IUPAC Name: N-{3-[3-(dimethylamino)prop-2-enoyl]phenyl}-N-ethylacetamide
Synonyms: N-{3-[3-(dimethylamino)prop-2-enoyl]phenyl}-N-ethylacetamide
Molecular Weight (g/mol): 260.34
Molecular Formula: C15H20N2O2
InChi Key: UXWJJVRASIHSQS-UHFFFAOYSA-N
SMILES: CCN(C(C)=O)C1=CC=CC(=C1)C(=O)C=CN(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results