missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyphenyl)thiourea, 97%

Catalog No. AAL1051903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1051903 1 g
AAL1051906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1051903 Supplier Thermo Scientific Chemicals Supplier No. L1051903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1520-26-9 | C7H8N2OS | 168.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1520-26-9
Molecular Formula C7H8N2OS
Molecular Weight (g/mol) 168.214
MDL Number MFCD00041159
InChI Key VYBCFZXLXJUFPM-UHFFFAOYSA-N
Synonym 1-2-hydroxyphenyl thiourea, 2-hydroxyphenyl thiourea, 2-hydroxy-phenyl-thiourea, 2-aminothioxomethyl amino phenol, o-hydroxyphenylthiourea, acmc-20ant0, thiourea, hydroxyphenyl, 1-o-hydroxyphenyl thiourea, n-2-hydroxyphenyl thiourea, thiourea, 2-hydroxyphenyl
PubChem CID 3003569
IUPAC Name (2-hydroxyphenyl)thiourea
SMILES C1=CC=C(C(=C1)NC(=S)N)O

Specifications

Melting Point ∽158°C (decomposition)
Odor Odorless
Quantity 1 g
Beilstein 1102479
Formula Weight 168.22
Percent Purity 97%
Chemical Name or Material N-(2-Hydroxyphenyl)thiourea
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only