accessibility menu, dialog, popup

UserName

CAS RN 1520-26-9

IUPAC Name: (2-hydroxyphenyl)thiourea
Synonyms: 2-hydroxyphenylthiourea
Molecular Weight (g/mol): 168.21
Molecular Formula: C7H8N2OS
InChi Key: VYBCFZXLXJUFPM-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results