missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxy-4-methylphenyl)acetamide, 97%, Thermo Scientific Chemicals

Catalog No. AAH66423MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66423MD 250 mg
AAH6642303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66423MD Supplier Thermo Scientific Chemicals Supplier No. H66423MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13429-10-2 | C9H11NO2 | 165.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13429-10-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00012061
InChI Key HTVPAACTHAQQAS-UHFFFAOYSA-N
Synonym n-2-hydroxy-4-methylphenyl acetamide, acetamide, n-2-hydroxy-4-methylphenyl, acetamide,n-2-hydroxy-4-methylphenyl, 6-acetamino-m-kresol, 2'-hydroxy-p-acetotoluidide, 2'-hydroxy-4'-methylacetoanilide, 2-acetamido-5-methylphenol, n-2-hydroxy-4-methyl-phenyl-acetamide
PubChem CID 83437
IUPAC Name N-(2-hydroxy-4-methylphenyl)acetamide
SMILES CC1=CC(=C(C=C1)NC(=O)C)O

Specifications

Quantity 250 mg
Beilstein 972595
Formula Weight 165.19
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-(2-Hydroxy-4-methylphenyl)acetamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only