accessibility menu, dialog, popup

UserName

CAS RN 13429-10-2

IUPAC Name: N-(2-hydroxy-4-methylphenyl)acetamide
Synonyms: N-(2-hydroxy-4-methylphenyl)acetamide
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: HTVPAACTHAQQAS-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C)C=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results