missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Diethylaminoethyl)-1-naphthylamine Oxalate 98.0+%, TCI America™
Supplier: TCI America D046925G
Specifications
| N-(2-Diethylaminoethyl)-1-naphthylamine Oxalate | |
| C18H24N2O4 | |
| 25 g | |
| MNUSPWMHIHYMKM-UHFFFAOYSA-N | |
| N-[2-(diethylamino)ethyl]naphthalen-1-amine; oxalic acid | |
| 44630228 | |
| ≥98.0% (T) |
| 29473-53-8 | |
| MFCD00060156 | |
| N,N-Diethyl-N′-1-naphthylethylenediamine Oxalate, Tsuda′s Reagent | |
| OC(=O)C(O)=O.CCN(CC)CCNC1=C2C=CC=CC2=CC=C1 | |
| 332.40 | |
| 332.40 | |
| Crystalline Powder |
Chemical Identifiers
| 29473-53-8 | |
| 332.40 | |
| MNUSPWMHIHYMKM-UHFFFAOYSA-N | |
| N-[2-(diethylamino)ethyl]naphthalen-1-amine; oxalic acid |
| C18H24N2O4 | |
| MFCD00060156 | |
| 44630228 | |
| OC(=O)C(O)=O.CCN(CC)CCNC1=C2C=CC=CC2=CC=C1 |
Safety and Handling
TSCA : No