accessibility menu, dialog, popup

UserName

CAS RN 29473-53-8

IUPAC Name: N-[2-(diethylamino)ethyl]naphthalen-1-amine; oxalic acid
Synonyms: N-[2-(diethylamino)ethyl]naphthalen-1-amine; oxalic acid
Molecular Weight (g/mol): 332.4
Molecular Formula: C18H24N2O4
InChi Key: MNUSPWMHIHYMKM-UHFFFAOYSA-N
SMILES: OC(=O)C(O)=O.CCN(CC)CCNC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results