missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Diethylaminoethyl)-1-naphthylamine Oxalate 98.0+%, TCI America™

Catalog No. p-7126232
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D04691G 1 g
D04695G 5 g
D046925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D04691G Supplier TCI America Supplier No. D04691G

Chemical Identifiers

CAS 29473-53-8
Molecular Formula C18H24N2O4
Molecular Weight (g/mol) 332.40
MDL Number MFCD00060156
InChI Key MNUSPWMHIHYMKM-UHFFFAOYSA-N
PubChem CID 44630228
IUPAC Name N-[2-(diethylamino)ethyl]naphthalen-1-amine; oxalic acid
SMILES OC(=O)C(O)=O.CCN(CC)CCNC1=C2C=CC=CC2=CC=C1

Specifications

Quantity 1 g
Synonym N,N-Diethyl-N′C-1-naphthylethylenediamine Oxalate, Tsuda′Cs Reagent
Formula Weight 332.40
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Diethylaminoethyl)-1-naphthylamine Oxalate