missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2'-Cyanobiphenyl-4-ylmethyl)-L-valine Methyl Ester Hydrochloride 98.0+%, TCI America™
$267.61 - $562.15
Chemical Identifiers
| CAS | 482577-59-3 |
|---|---|
| Molecular Formula | C20H23ClN2O2 |
| Molecular Weight (g/mol) | 358.866 |
| MDL Number | MFCD08460094 |
| InChI Key | AZQXUWUZQLZNIM-FYZYNONXSA-N |
| PubChem CID | 16744793 |
| IUPAC Name | methyl (2S)-2-[[4-(2-cyanophenyl)phenyl]methylamino]-3-methylbutanoate;hydrochloride |
| SMILES | CC(C)C(C(=O)OC)NCC1=CC=C(C=C1)C2=CC=CC=C2C#N.Cl |
Chemical Identifiers
| 482577-59-3 | |
| 358.866 | |
| AZQXUWUZQLZNIM-FYZYNONXSA-N | |
| methyl (2S)-2-[[4-(2-cyanophenyl)phenyl]methylamino]-3-methylbutanoate;hydrochloride |
| C20H23ClN2O2 | |
| MFCD08460094 | |
| 16744793 | |
| CC(C)C(C(=O)OC)NCC1=CC=C(C=C1)C2=CC=CC=C2C#N.Cl |
Specifications
| 482577-59-3 | |
| White | |
| MFCD08460094 | |
| AZQXUWUZQLZNIM-FYZYNONXSA-N | |
| methyl (2S)-2-[[4-(2-cyanophenyl)phenyl]methylamino]-3-methylbutanoate;hydrochloride | |
| 16744793 | |
| ≥98.0% (HPLC,T) | |
| N-(2′-Cyanobiphenyl-4-ylmethyl)-L-valine Methyl Ester Hydrochloride |
| 187°C | |
| C20H23ClN2O2 | |
| 5 g | |
| CC(C)C(C(=O)OC)NCC1=CC=C(C=C1)C2=CC=CC=C2C#N.Cl | |
| 358.866 | |
| 358.87 | |
| Crystalline Powder |
Safety and Handling
TSCA : No