CAS RN 482577-59-3
CAS RN 482577-59-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(2'-Cyanobiphenyl-4-ylmethyl)-L-valine Methyl Ester Hydrochloride 98.0+%, TCI America™
CAS: 482577-59-3 Molecular Formula: C20H23ClN2O2 Molecular Weight (g/mol): 358.866 MDL Number: MFCD08460094 InChI Key: AZQXUWUZQLZNIM-FYZYNONXSA-N PubChem CID: 16744793 IUPAC Name: methyl (2S)-2-[[4-(2-cyanophenyl)phenyl]methylamino]-3-methylbutanoate;hydrochloride SMILES: CC(C)C(C(=O)OC)NCC1=CC=C(C=C1)C2=CC=CC=C2C#N.Cl