missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Dinitrophenyl)-L-serine 98.0+%, TCI America™

Catalog No. D1037100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1037100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1037100MG Supplier TCI America Supplier No. D1037100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1655-64-7
Molecular Formula C9H9N3O7
Molecular Weight (g/mol) 271.185
MDL Number MFCD00038098
InChI Key SBQZBOCQYMVLTC-ZETCQYMHSA-N
Synonym n-2,4-dinitrophenyl-l-serine, dnp-ser, n-dnp-l-serine, 2s-2-2,4-dinitrophenyl amino-3-hydroxypropanoic acid, 1oau, serine, n-2,4-dinitrophenyl-, dl, dnp-ser-oh, ser-dnf, n-2-4-dnp-l-serine, n-2,4-dnp-l-serine
PubChem CID 96800
ChEBI CHEBI:53084
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])NC(CO)C(=O)O

Specifications

Melting Point 177°C
Color Yellow
Quantity 100 mg
UN Number 1325
Formula Weight 271.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2,4-Dinitrophenyl)-L-serine