CAS RN 1655-64-7
IUPAC Name:
2-[(2,4-dinitrophenyl)amino]-3-hydroxypropanoic acid
Synonyms:
N-(2,4-dinitrophenyl)serine
Molecular Weight (g/mol):
271.19
Molecular Formula:
C9H9N3O7
InChi Key:
SBQZBOCQYMVLTC-UHFFFAOYNA-N
SMILES:
OCC(NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O
1
–
1
of
1
results
1
N-(2,4-Dinitrophenyl)-L-serine 98.0+%, TCI America™
CAS: 1655-64-7 Molecular Formula: C9H9N3O7 Molecular Weight (g/mol): 271.185 MDL Number: MFCD00038098 InChI Key: SBQZBOCQYMVLTC-ZETCQYMHSA-N Synonym: n-2,4-dinitrophenyl-l-serine,dnp-ser,n-dnp-l-serine,2s-2-2,4-dinitrophenyl amino-3-hydroxypropanoic acid,1oau,serine, n-2,4-dinitrophenyl-, dl,dnp-ser-oh,ser-dnf,n-2-4-dnp-l-serine,n-2,4-dnp-l-serine PubChem CID: 96800 ChEBI: CHEBI:53084 SMILES: C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])NC(CO)C(=O)O