missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Dinitrophenyl)glycine 98.0+%, TCI America™

Catalog No. D1032100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1032100MG 100 mg
D10321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1032100MG Supplier TCI America Supplier No. D1032100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1084-76-0
Molecular Formula C8H7N3O6
Molecular Weight (g/mol) 241.159
MDL Number MFCD00024400
InChI Key RQPREKYEHBAOAR-UHFFFAOYSA-N
Synonym N-Dnp-glycine, Dnp-Gly-OH
PubChem CID 14135
IUPAC Name 2-(2,4-dinitroanilino)acetic acid
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])NCC(=O)O

Specifications

Melting Point 200°C
Color Yellow
Quantity 100 mg
UN Number 1325
Formula Weight 241.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2,4-Dinitrophenyl)glycine
RTECSNumber MC0070000
TSCA No