accessibility menu, dialog, popup

UserName

CAS RN 1084-76-0

IUPAC Name: 2-[(2,4-dinitrophenyl)amino]acetic acid
Synonyms: [(2,4-dinitrophenyl)amino]acetic acid
Molecular Weight (g/mol): 241.16
Molecular Formula: C8H7N3O6
InChi Key: RQPREKYEHBAOAR-UHFFFAOYSA-N
SMILES: OC(=O)CNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results