missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2,4-Dinitrophenyl)-6-aminohexanoic Acid 95.0+%, TCI America™
$57.79 - $281.48
Chemical Identifiers
| CAS | 10466-72-5 |
|---|---|
| Molecular Formula | C12H15N3O6 |
| Molecular Weight (g/mol) | 297.27 |
| MDL Number | MFCD00038470 |
| InChI Key | ZYUWUKIAUDIXCQ-UHFFFAOYSA-N |
| Synonym | N-Dnp-6-aminohexanoic Acid |
| PubChem CID | 96812 |
| ChEBI | CHEBI:53698 |
| IUPAC Name | 6-[(2,4-dinitrophenyl)amino]hexanoic acid |
| SMILES | OC(=O)CCCCCNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D2254100MG
|
TCI America
D2254100MG |
100 mg |
Each for $57.79
|
|
|||||
|
D22541G
|
TCI America
D22541G |
1 g |
Each for $281.48
|
|
|||||
Chemical Identifiers
| 10466-72-5 | |
| 297.27 | |
| ZYUWUKIAUDIXCQ-UHFFFAOYSA-N | |
| 96812 | |
| 6-[(2,4-dinitrophenyl)amino]hexanoic acid |
| C12H15N3O6 | |
| MFCD00038470 | |
| N-Dnp-6-aminohexanoic Acid | |
| CHEBI:53698 | |
| OC(=O)CCCCCNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O |
Specifications
| 10466-72-5 | |
| Yellow | |
| MFCD00038470 | |
| N-Dnp-6-aminohexanoic Acid | |
| OC(=O)CCCCCNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O | |
| 297.27 | |
| CHEBI:53698 | |
| ≥95.0% (HPLC,T) | |
| N-(2,4-Dinitrophenyl)-6-aminohexanoic Acid |
| 133°C | |
| C12H15N3O6 | |
| 100 mg | |
| ZYUWUKIAUDIXCQ-UHFFFAOYSA-N | |
| 6-[(2,4-dinitrophenyl)amino]hexanoic acid | |
| 96812 | |
| 297.27 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer