missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Dinitrophenyl)-6-aminohexanoic Acid 95.0+%, TCI America™

Supplier:  TCI America D2254100MG

 View more versions of this product

Catalog No. D2254100MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-(2,4-Dinitrophenyl)-6-aminohexanoic Acid
133°C
C12H15N3O6
100 mg
ZYUWUKIAUDIXCQ-UHFFFAOYSA-N
6-[(2,4-dinitrophenyl)amino]hexanoic acid
96812
297.27
Crystalline Powder
10466-72-5
Yellow
MFCD00038470
N-Dnp-6-aminohexanoic Acid
OC(=O)CCCCCNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
297.27
CHEBI:53698
≥95.0% (HPLC,T)

Chemical Identifiers

10466-72-5
297.27
ZYUWUKIAUDIXCQ-UHFFFAOYSA-N
96812
6-[(2,4-dinitrophenyl)amino]hexanoic acid
C12H15N3O6
MFCD00038470
N-Dnp-6-aminohexanoic Acid
CHEBI:53698
OC(=O)CCCCCNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions