missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Myricetin | |
| 360°C | |
| MFCD00006827 | |
| myricetin, cannabiscetin, myricetol, myricitin, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-chromen-4-one, 3,3',4',5,5',7-hexahydroxyflavone, 3,5,7,3',4',5'-hexahydroxyflavone, unii-76xc01ftoj, ccris 5838, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-1-benzopyran-4-one | |
| OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C(O)=C2)=C1 | |
| 318.24 | |
| CHEBI:18152 | |
| ≥97.0% (HPLC) |
| 529-44-2 | |
| C15H10O8 | |
| 5 g | |
| IKMDFBPHZNJCSN-UHFFFAOYSA-N | |
| 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one | |
| 5281672 | |
| 318.24 | |
| Crystalline Powder |
Chemical Identifiers
| 529-44-2 | |
| 318.24 | |
| IKMDFBPHZNJCSN-UHFFFAOYSA-N | |
| 5281672 | |
| 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one |
| C15H10O8 | |
| MFCD00006827 | |
| myricetin, cannabiscetin, myricetol, myricitin, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-chromen-4-one, 3,3',4',5,5',7-hexahydroxyflavone, 3,5,7,3',4',5'-hexahydroxyflavone, unii-76xc01ftoj, ccris 5838, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-1-benzopyran-4-one | |
| CHEBI:18152 | |
| OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C(O)=C2)=C1 |
Safety and Handling
RTECSNumber : LK8646000
TSCA : No