accessibility menu, dialog, popup

UserName

CAS RN 529-44-2

IUPAC Name: 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one
Synonyms: myricetin MYC myricitin cannabiscetin myricetol
Molecular Weight (g/mol): 318.24
Molecular Formula: C15H10O8
InChi Key: IKMDFBPHZNJCSN-UHFFFAOYSA-N
SMILES: OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C(O)=C2)=C1
Percent Purity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)