missing translation for 'onlineSavingsMsg'
Learn More

Myricetin 97.0+%, TCI America™

Catalog No. p-7133041
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2131500MG 500 mg
M21315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2131500MG Supplier TCI America Supplier No. M2131500MG
Only null left

Chemical Identifiers

CAS 529-44-2
Molecular Formula C15H10O8
Molecular Weight (g/mol) 318.24
MDL Number MFCD00006827
InChI Key IKMDFBPHZNJCSN-UHFFFAOYSA-N
Synonym myricetin, cannabiscetin, myricetol, myricitin, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-chromen-4-one, 3,3',4',5,5',7-hexahydroxyflavone, 3,5,7,3',4',5'-hexahydroxyflavone, unii-76xc01ftoj, ccris 5838, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-1-benzopyran-4-one
PubChem CID 5281672
ChEBI CHEBI:18152
IUPAC Name 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one
SMILES OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C(O)=C2)=C1

Specifications

Melting Point 360°C
Quantity 500 mg
Formula Weight 318.24
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Myricetin
RTECSNumber LK8646000
TSCA No