missing translation for 'onlineSavingsMsg'
Learn More

Myricetin 97.0+%, TCI America™

Supplier:  TCI America M2131500MG

 View more versions of this product

Catalog No. M2131500MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Myricetin
360°C
MFCD00006827
myricetin, cannabiscetin, myricetol, myricitin, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-chromen-4-one, 3,3',4',5,5',7-hexahydroxyflavone, 3,5,7,3',4',5'-hexahydroxyflavone, unii-76xc01ftoj, ccris 5838, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-1-benzopyran-4-one
OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C(O)=C2)=C1
318.24
CHEBI:18152
≥97.0% (HPLC)
529-44-2
C15H10O8
500 mg
IKMDFBPHZNJCSN-UHFFFAOYSA-N
3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one
5281672
318.24
Crystalline Powder

Chemical Identifiers

529-44-2
318.24
IKMDFBPHZNJCSN-UHFFFAOYSA-N
5281672
3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one
C15H10O8
MFCD00006827
myricetin, cannabiscetin, myricetol, myricitin, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-chromen-4-one, 3,3',4',5,5',7-hexahydroxyflavone, 3,5,7,3',4',5'-hexahydroxyflavone, unii-76xc01ftoj, ccris 5838, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-1-benzopyran-4-one
CHEBI:18152
OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C(O)=C2)=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : LK8646000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions