missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MYOD1 Primer

Catalog No. 361062306
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361062306 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361062306 Supplier Novus Biologicals™ Supplier No. NBP1716520.5ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Content And Storage -20°C. Avoid freeze/thaw cycles.
Quantity 0.5 mL
For Use With (Application) ChIP
Type Primer