missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals myo-Inositol, Cell Culture Grade

A lipotropic agent | CAS: 87-89-8 | C6H12O6 | 180.156 g/mol

Supplier:  Thermo Scientific Chemicals J6288622

 View more versions of this product

Catalog No. AAJ6288622

Only null left
Explore available promotions

Description

Description

It has been tested with CHO, L929, and Vero cell lines and can be used for other cell culture applications. A method for HPLC identification and quantitation has been achieved by this chemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been tested with CHO, L929, and Vero cell lines and can be used for other cell culture applications. A method for HPLC identification and quantitation has been achieved by this chemical.

Solubility
It is soluble in water (50 mg/ml).

Notes
Solutions of myo-inositol can be autoclaved for use in cell culture. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

myo-Inositol
222°C to 227°C
Odorless
MFCD00077932
1907329
scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
CDAISMWEOUEBRE-UHFFFAOYSA-N
cyclohexane-1,2,3,4,5,6-hexol
892
180.16
Crystalline Powder
87-89-8
White
C6H12O6
100 g
14,4978
It is soluble in water (50mg/ml).
C1(C(C(C(C(C1O)O)O)O)O)O
180.156
CHEBI:24848
Cell Culture

Chemical Identifiers

87-89-8
180.156
CDAISMWEOUEBRE-UHFFFAOYSA-N
892
cyclohexane-1,2,3,4,5,6-hexol
C6H12O6
MFCD00077932
scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
CHEBI:24848
C1(C(C(C(C(C1O)O)O)O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-781-2

RTECSNumber : NM7520800

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only