missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals myo-Inositol, Cell Culture Grade

Catalog No. p-7042544
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6288618 50 g
AAJ6288622 100 g
AAJ6288630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6288618 Supplier Thermo Scientific Chemicals Supplier No. J6288618

A lipotropic agent | CAS: 87-89-8 | C6H12O6 | 180.156 g/mol

It has been tested with CHO, L929, and Vero cell lines and can be used for other cell culture applications. A method for HPLC identification and quantitation has been achieved by this chemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been tested with CHO, L929, and Vero cell lines and can be used for other cell culture applications. A method for HPLC identification and quantitation has been achieved by this chemical.

Solubility
It is soluble in water (50 mg/ml).

Notes
Solutions of myo-inositol can be autoclaved for use in cell culture. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00077932
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
Synonym scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
PubChem CID 892
ChEBI CHEBI:24848
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES C1(C(C(C(C(C1O)O)O)O)O)O

Specifications

Melting Point 222°C to 227°C
Color White
Odor Odorless
Quantity 50 g
Beilstein 1907329
Merck Index 14,4978
Solubility Information It is soluble in water (50mg/ml).
Formula Weight 180.16
Grade Cell Culture
Physical Form Crystalline Powder
Chemical Name or Material myo-Inositol
Show More Show Less
EINECSNumber 201-781-2
RTECSNumber NM7520800
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only