missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MycoSEQ™ Mycoplasma Detection Kits
GREENER_CHOICE

Catalog No. 4460626
Change view
Click to view available options
Product Type:
Assay Kit
Assay Kit and Sample Preparation
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
4460626 Assay Kit and Sample Preparation
4460623 Assay Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4460626 Supplier Applied Biosystems™ Supplier No. 4460626*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The MycoSEQ Mycoplasma Detection Kit is a real-time mycoplasma PCR assay designed and validated to meet the requirements of European Pharmacopeia Chapter 2.6.7 for use as a nucleic acid amplification technique (NAT).

The MycoSEQ Mycoplasma Detection Kit is a real-time mycoplasma PCR assay designed and validated to meet the requirements of European Pharmacopeia Chapter 2.6.7 for use as a nucleic acid amplification technique (NAT). The PCR primers were designed for mycoplasma detection using a proprietary bioinformatics pipeline to detect >90 mycoplasma species with high specificity and no cross-reactivity to closely-related bacterial species. The assay's sensitivity has been demonstrated by internal and external validations to detect 10 CFU/mL or the genomic equivalent as recommended by regulatory guidance.

Features of the MycoSEQ Mycoplasma Detection Kit include:

  • Easy to use, results in less than one day
  • Includes a unique discriminatory positive control to differentiate a true positive result from a potential cross-contamination positive call
  • Includes protocols for various methods, including lot release and in-process testing
  • Live mycoplasma is not needed: MycoSEQ Mycoplasma Detection Kit does not contain any live mycoplasma and does not require use of live mycoplasma for validation
  • Can be used with gDNA, inactivated mycoplasma, or live mycoplasma
  • May be used as an alternative to the standard 28-day culture test
  • Can be used in conjunction with culture-based methods by providing preliminary results while waiting for results from the 28-day test

The PrepSEQ nucleic acid extraction module included in MycoSEQ Mycoplasma Detection Kit, with PrepSEQ sample preparation (Cat. No. 4460626) offers a robust and easy workflow to overcome complex matrix challenges to ensure the required sensitivity. For other recommended sample prep options, see the Integrated workflow solution section.

Integrated workflow solution

The MycoSEQ Mycoplasma Detection Kit is part of an integrated workflow for adventitious agent, impurity and contaminant testing during biopharmaceutical manufacturing. The entire workflow, including PrepSEQ Sample Preparation Kits together with qPCR analysis, can provide results in under 5 hours. (Please note complex matrices may require additional upfront processing steps, as described in various available protocols.)

MycoSEQ kit performance has been tested and confirmed on the Applied Biosystems 7500 Fast Real-Time PCR System and the Applied Biosystems QuantStudio 5 Real-Time PCR System. Data analysis using AccuSEQ Real-Time PCR Detection Software provides accurate quantitation and security, audit, and e-signature capabilities to help enable 21 CFR Pt 11 compliance.

Specifications

Content And Storage MycoSEQ Mycoplasma Detection Kit
• Power SYBR Green PCR Master Mix (2X), store at -15°C to -25°C on receipt, 2°C to 8°C after first use, protected from light
• Mycoplasma Real-Time PCR Primer Mix (10X), -15°C to -25°C on receipt, 2°C to 8°C after first use
• Discriminatory Positive/Extraction Control, -15°C to -25°C
• Negative control, -15°C to -25°C on receipt, 2°C to 8°C after first use

PrepSEQ 1-2-3 Mycoplasma Nucleic Acid Sample Extraction Kit
• Cell Fractionation Buffer, store at 2°C to 8°C
• RNase Cocktail Enzyme Mix, -15°C to -25°C
• Lysis Buffer, 18°C to 25°C
• Wash Buffer Concentrate, 18°C to 25°C
• Elution Buffer, 18°C to 25°C
• Proteinase K Buffer, 18°C to 25°C
• Magnetic Particles, 18°C to 25°C
• Proteinase K, -15°C to -25°C

Description Detection for BioProduction process media, cells, raw materials
For Use With QS5 0.1 mL Block or 7500 FAST
Label or Dye SYBR Green
Product Line MycoSEQ
Product Type Assay Kit and Sample Preparation
Quantity 100 Reactions
For Use With (Application) Mycoplasma Detection
How can I remove mycoplasma contamination from my cell culture medium?

Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.