missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Murexide, TCI America™
$45.57 - $81.64
Chemical Identifiers
| CAS | 3051-09-0 |
|---|---|
| Molecular Formula | C8H3N5O6-2 |
| Molecular Weight (g/mol) | 265.141 |
| MDL Number | MFCD00012777 |
| InChI Key | MUMMBXDKKJEASE-UHFFFAOYSA-L |
| Synonym | Ammonium Purpurate |
| PubChem CID | 51381221 |
| IUPAC Name | 6-oxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1H-pyrimidine-2,4-diolate |
| SMILES | C1(=C(N=C(NC1=O)[O-])[O-])N=C2C(=O)NC(=O)NC2=O |
Chemical Identifiers
| 3051-09-0 | |
| 265.141 | |
| MUMMBXDKKJEASE-UHFFFAOYSA-L | |
| 51381221 | |
| C1(=C(N=C(NC1=O)[O-])[O-])N=C2C(=O)NC(=O)NC2=O |
| C8H3N5O6-2 | |
| MFCD00012777 | |
| Ammonium Purpurate | |
| 6-oxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1H-pyrimidine-2,4-diolate |
Specifications
| 3051-09-0 | |
| MFCD00012777 | |
| MUMMBXDKKJEASE-UHFFFAOYSA-L | |
| 6-oxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1H-pyrimidine-2,4-diolate | |
| 51381221 | |
| Purple/Red | |
| Murexide |
| C8H3N5O6-2 | |
| Ammonium Purpurate | |
| C1(=C(N=C(NC1=O)[O-])[O-])N=C2C(=O)NC(=O)NC2=O | |
| 265.141 | |
| 284.19 | |
| Crystal/Powder |
Safety and Handling
TSCA : Yes