missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Murexide Indicator, 0.2% (w/w) in Sodium Chloride, Powder Mixture, For Calcium, APHA, Spectrum™ Chemical
$500.75 - $1,707.50
Specifications
| CAS | 3051-09-0 , 7647-14-5 |
|---|---|
| Concentration | 0.2 Percent (w/w) |
| Grade | APHA |
| Packaging | Poly Bottle |
| Color | Purple/Red to Red |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.