missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Murexide (Ammonium Purpurate), Reagents
Murexide (Ammonium Purpurate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$128.43 - $405.82
Chemical Identifiers
| CAS | 3051-09-0 |
|---|---|
| Molecular Formula | C8H8N6O6 |
| Molecular Weight (g/mol) | 284.19 |
| InChI Key | PCCZWUCAYILEMX-UHFFFAOYSA-O |
| Synonym | Ammonium Purpurate, Color Index 56085 |
| IUPAC Name | ammonium 2,4,6-trioxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1,3-diazinan-5-ide |
| SMILES | [NH4+].O=C1NC(=O)[C-](N=C2C(=O)NC(=O)NC2=O)C(=O)N1 |
Chemical Identifiers
| 3051-09-0 | |
| 284.19 | |
| Ammonium Purpurate, Color Index 56085 | |
| [NH4+].O=C1NC(=O)[C-](N=C2C(=O)NC(=O)NC2=O)C(=O)N1 |
| C8H8N6O6 | |
| PCCZWUCAYILEMX-UHFFFAOYSA-O | |
| ammonium 2,4,6-trioxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1,3-diazinan-5-ide |
Specifications
| >300°C | |
| 1500°C | |
| Ammonium Purpurate, Color Index 56085 | |
| [NH4+].O=C1NC(=O)[C-](N=C2C(=O)NC(=O)NC2=O)C(=O)N1 | |
| 284.19 | |
| 1 | |
| Purple/Red to Red | |
| Murexide (Ammonium Purpurate) |
| 3051-09-0 | |
| C8H8N6O6 | |
| PCCZWUCAYILEMX-UHFFFAOYSA-O | |
| ammonium 2,4,6-trioxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1,3-diazinan-5-ide | |
| Glass Bottle | |
| Pink (Excess Calcium) to Purple (Excess EDTA) | |
| Solid |