missing translation for 'onlineSavingsMsg'
Learn More

Murexide (Ammonium Purpurate), Reagents

Catalog No. 01337166
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337166 1 g
01337165 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337166 Supplier Labchem Inc Supplier No. C22519801C2
Only null left

Murexide (Ammonium Purpurate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 3051-09-0
Molecular Formula C8H8N6O6
Molecular Weight (g/mol) 284.19
InChI Key PCCZWUCAYILEMX-UHFFFAOYSA-O
Synonym Ammonium Purpurate, Color Index 56085
IUPAC Name ammonium 2,4,6-trioxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1,3-diazinan-5-ide
SMILES [NH4+].O=C1NC(=O)[C-](N=C2C(=O)NC(=O)NC2=O)C(=O)N1

Specifications

Melting Point >300°C
Boiling Point 1500°C
Packaging Glass Bottle
Specific Gravity 1
pH Color Changes Pink (Excess Calcium) to Purple (Excess EDTA)
Color Purple/Red to Red
Quantity 1 g
Physical Form Solid
Chemical Name or Material Murexide (Ammonium Purpurate)