Learn More
MP Biomedicals™ Murashige and Skoog Media
All Murashige and Skoog plant cell culture Media are manufactured from high quality chemicals and reagents.
Supplier: MP Biomedicals™ 092623122
Description
- Each formulation is prepared according to the original specification, except where noted
- Finished lots of media are plant cell tested
Specifications
| Powder | |
| 1 x 10 L | |
| 3.75 to 4.75 |
| Murashige and Skoog Basal Medium | |
| Solution | |
| 4°C |