missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Murashige and Skoog Media
All Murashige and Skoog plant cell culture Media are manufactured from high quality chemicals and reagents.
Supplier: MP Biomedicals™ 092610020
Description
- Each formulation is prepared according to the original specification, except where noted
- Finished lots of media are plant cell tested
Specifications
| Powder | |
| No Sucrose, No Agar, No IAA, No Kinetin | |
| Solution | |
| 4°C |
| Murashige and Skoog Medium | |
| 10 x 1 L | |
| 4.80 to 5.65 |