missing translation for 'onlineSavingsMsg'
Learn More

Multi-Element Organometallic Oil Standards 23 Element, 500μg/g; SPEX CertiPrep™

Catalog No. S23500Z
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S23500Z 50 g
S23500Y 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S23500Z Supplier SPEX CertiPrep Group LLC Supplier No. S23500Z
Only null left

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag500μg/g
Al500μg/g
B500μg/g
Ba500μg/g
Ca500μg/g
Cd500μg/g
Cr500μg/g
Cu500μg/g
Fe500μg/g
K500μg/g
Mg500μg/g
Mn500μg/g
Mo500μg/g
Na500μg/g
Ni500μg/g
P500μg/g
Pb500μg/g
Sb500μg/g
Si500μg/g
Sn500μg/g
Ti500μg/g
V500μg/g
Zn500μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material 23 Element Oil Standard
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Analyte or Component Names Al,Ba,B
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Element Al, Ba, B
Solvent or Matrix Base Oil 75
Industry Type Petrochemical Testing
Quantity 50 g

Product intended for laboratory use only