missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Multi-Element Organometallic Oil Standards 23 Element, 300μg/g; SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$1,332.74 - $1,332.74
Specifications
| Chemical Name or Material | 23 Element Oil Standard |
|---|---|
| Packaging | Triple-leached LDPE bottle |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| Analyte or Component Names | Al,Ba,B |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ag | 300μg/g |
| Al | 300μg/g |
| B | 300μg/g |
| Ba | 300μg/g |
| Ca | 300μg/g |
| Cd | 300μg/g |
| Cr | 300μg/g |
| Cu | 300μg/g |
| Fe | 300μg/g |
| K | 300μg/g |
| Mg | 300μg/g |
| Mn | 300μg/g |
| Mo | 300μg/g |
| Na | 300μg/g |
| Ni | 300μg/g |
| P | 300μg/g |
| Pb | 300μg/g |
| Sb | 300μg/g |
| Si | 300μg/g |
| Sn | 300μg/g |
| Ti | 300μg/g |
| V | 300μg/g |
| Zn | 300μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| 23 Element Oil Standard | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Al, Ba, B | |
| Petrochemical Testing |
| Triple-leached LDPE bottle | |
| Al,Ba,B | |
| Certified Reference Material | |
| Base Oil 75 |
Product intended for laboratory use only