Learn More
Gilson™ PIPETMAN™ M Multi-Channel
GREENER_CHOICE
PIPETMAN™ M Multi-Channel is a Bluetooth™-enabled electronic pipette that offers accuracy and precision in both standard and repetitive pipetting modes.
Supplier:  Gilson™ F81050
 View more versions of this product
Catalog No. F81050G
Get Up to $1,500 in Gilson Products with Purchase
View all promotions
Only null left
Description
Description
  • Allows accurate and precise pipetting
  • The motorized piston allows for high pipetting reproducibility and accuracy in all modes
  • Associated with TRACKMAN™ Connected and its PipettePilot™ application, PIPETMAN™ M Connected reduces the risk of human error by transferring verifiable pipetting parameters and commands in the pipette
  • Increased functionality
  • The intuitive interface has five pipetting modes for a large number of applications: pipette, repetitive, mix, reverse, and custom
  • Create and transfer up to 10 custom protocols from PIPETMAN™ M Connected Software into your pipette
  • Bluetooth™ connection enables communication with TRACKMAN™ Connected and helps guide you through your protocol telling you where, when, and how much to pipette with real-time interactions
  • Simple and comfortable to use
  • Aspirate and dispense with one click of the push button and eject tips with ease
  • The lightweight and ergonomic design rests comfortably in your hand for maximum pipetting comfort
Specifications
Chemical Identifiers
Universal
Autoclavable
Pipette Tip, TRACKMAN™ Connected
Polypropylene
Yes
Pipette
1
15.75 in.
5.9 in.
2.16 in.
3.7 V and 420 mAh (1.55 Wh)
No
Reusable
Ergonomic
Pipetman M
Non-sterile
Pipette Tips, TRACKMAN™ Connected
1 Year
32.4 cm
15 cm
5.5 cm
Certified Quality
Specifications
Universal
Autoclavable
Reusable
Ergonomic
P8x20M BT
Pipetman M
Non-sterile
1 to 20 μL
1
15.75 in.
5.9 in.
2.16 in.
3.7 V and 420 mAh (1.55 Wh)
No
Yellow
Pipette Tip, TRACKMAN™ Connected
Polypropylene
8
Yes
Pipette
Pipette Tips, TRACKMAN™ Connected
1 Year
32.4 cm
15 cm
5.5 cm
Certified Quality
Product Suggestions
Product Suggestions