missing translation for 'onlineSavingsMsg'
Learn More

Mucic Acid 97.0+%, TCI America™

$60.75 - $225.93

Chemical Identifiers

CAS 526-99-8
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.138
MDL Number MFCD00004239
InChI Key DSLZVSRJTYRBFB-DUHBMQHGSA-N
Synonym galactaric acid, mucic acid, 2r,3s,4r,5s-2,3,4,5-tetrahydroxyhexanedioic acid, saccharolactic acid, mucicacid, unii-e149j5otif, d-galactaric acid, galactosaccharic acid, galactarate, meso-galactaric acid
PubChem CID 3037582
ChEBI CHEBI:30852
IUPAC Name (2S,3R,4S,5R)-2,3,4,5-tetrahydroxyhexanedioic acid
SMILES C(C(C(C(=O)O)O)O)(C(C(=O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
M046625G
View Documents Promotion Details
N/A 25 g
Each for $60.75
Only null left
 
M0466500G
View Documents Promotion Details
N/A 500 g
Each for $225.93
Only null left
 
Description

Description

Specifications

Chemical Identifiers

526-99-8
210.138
DSLZVSRJTYRBFB-DUHBMQHGSA-N
3037582
(2S,3R,4S,5R)-2,3,4,5-tetrahydroxyhexanedioic acid
C6H10O8
MFCD00004239
galactaric acid, mucic acid, 2r,3s,4r,5s-2,3,4,5-tetrahydroxyhexanedioic acid, saccharolactic acid, mucicacid, unii-e149j5otif, d-galactaric acid, galactosaccharic acid, galactarate, meso-galactaric acid
CHEBI:30852
C(C(C(C(=O)O)O)O)(C(C(=O)O)O)O

Specifications

526-99-8
White-Yellow
MFCD00004239
DSLZVSRJTYRBFB-DUHBMQHGSA-N
(2S,3R,4S,5R)-2,3,4,5-tetrahydroxyhexanedioic acid
3037582
210.14
Crystalline Powder
210°C
C6H10O8
galactaric acid, mucic acid, 2r,3s,4r,5s-2,3,4,5-tetrahydroxyhexanedioic acid, saccharolactic acid, mucicacid, unii-e149j5otif, d-galactaric acid, galactosaccharic acid, galactarate, meso-galactaric acid
C(C(C(C(=O)O)O)O)(C(C(=O)O)O)O
210.138
CHEBI:30852
≥97.0% (T)
Mucic Acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3410

RTECSNumber : LW5180000

TSCA : Yes

SDS
Documents

Documents