accessibility menu, dialog, popup

UserName

OHOHOHHOOOHOHO

CAS RN 526-99-8

IUPAC Name: 2,3,4,5-tetrahydroxyhexanedioic acid
Synonyms: D-saccharate altraric acid, D- potassium hydrogen glucarate altraric acid
Molecular Weight (g/mol): 210.14
Molecular Formula: C6H10O8
InChi Key: DSLZVSRJTYRBFB-UHFFFAOYNA-N
SMILES: OC(C(O)C(O)C(O)=O)C(O)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)