missing translation for 'onlineSavingsMsg'
Learn More

Mucic Acid 97.0+%, TCI America™

Catalog No. M0466500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0466500G 500 g
M046625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0466500G Supplier TCI America Supplier No. M0466500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 526-99-8
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.138
MDL Number MFCD00004239
InChI Key DSLZVSRJTYRBFB-DUHBMQHGSA-N
Synonym galactaric acid, mucic acid, 2r,3s,4r,5s-2,3,4,5-tetrahydroxyhexanedioic acid, saccharolactic acid, mucicacid, unii-e149j5otif, d-galactaric acid, galactosaccharic acid, galactarate, meso-galactaric acid
PubChem CID 3037582
ChEBI CHEBI:30852
IUPAC Name (2S,3R,4S,5R)-2,3,4,5-tetrahydroxyhexanedioic acid
SMILES C(C(C(C(=O)O)O)O)(C(C(=O)O)O)O

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 500 g
Formula Weight 210.14
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Mucic Acid
EINECSNumber (2)-3410
RTECSNumber LW5180000
TSCA Yes