missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MS(PEG)12 Methyl-PEG-NHS-Ester Reagent

Catalog No. PI22686
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI22686 1 g
PI22685 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI22686 Supplier Thermo Scientific™ Supplier No. LSG22686

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pegylate amines (-NH2 groups) on proteins (lysine residues) or assay surfaces using MS(PEG)n, amine-reactive (NHS ester), methyl-capped PEG compounds (4 to 24 EG units).

Thermo Scientific™ Pierce MS(PEG)n reagents are methyl-terminated, polyethylene glycol compounds (n equals 4 to 24 PEG units) activated as NHS esters for covalent pegylation of primary amines on proteins (e.g., lysines) or assay surfaces.

MS(PEG)n is the abbreviation for a set of compounds having polyethylene glycol (PEG) spacers with methyl (—CH3) and amine-reactive NHS-ester groups at opposite ends. The unbranched, hydrophilic, discrete-length molecules have the form Methyl-PEGn-NHS Ester, where the subscript “n” denotes 4, 8, 12, or 24 ethylene glycol units. The N-hydroxysuccinimide (NHS) ester is spontaneously reactive with primary amines (—NH2), providing for efficient PEGylation of proteins, peptides and other amine-containing molecules or surfaces.

Highlights:

  • NHS-activated for efficient PEGylation of primary amines at pH 7-9; reaction of NHS-ester group results in formation of stable, irreversible amide bonds
  • Fully characterized PEGylation reagents with defined PEG chain lengths; molecules of discrete molecular weight for consistency of performance in protein-modification applications
  • Provided as a series of 4, 8, 12 and 24 ethylene glycol units, enabling modification procedures to be optimized for a specific application while retaining all the benefits associated with protein PEGylation
  • PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
  • Easy-to-follow instructions increase the likelihood of a successful outcome

Specifications

Chemical Reactivity Amine
Content And Storage Store desiccated at -20°C.
Label Type Polyethylene Glycol (PEG), Blocking Groups
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Spacer 12 ethylene glycol units
Form Liquid
Quantity 1 g
Solubility Water
Product Type NHS-Ester Reagent
Can you provide the shelf-life for MS(PEG)12 Methyl-PEG-NHS-Ester Reagent?

MS(PEG)12 Methyl-PEG-NHS-Ester Reagent is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.