Learn More
Promega MS Compatible Yeast Protein Extract, Digest
Description
- Protein Extracts for Easier Mass Spec Instrument Monitoring
- Choose human or yeast, digested or intact extract
- Monitor LC and MS instrument performance
- Use for method development and optimization
Specifications
Specifications
| Content And Storage | -30°C to -10°C |
| Quantity | 100 μg |