missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MS 21570
Description
MS 21570 is a GPR171 antagonist (IC50 = 220 nM). Inhibits BigLEN (Cat.No. 6304) mediated increases in [35S]GTPγS binding in hypothalamic membranes and hyperpolarization of basolateral amygdala pyramidal neurons. Reduces anxiety-like behavior and fear conditioning in mice.
Specifications
Specifications
| Chemical Name or Material | N-Methyl-5-[(phenylmethyl)thio]-1,3,4-thiazadiazol-2-amine |
| CAS | 65373-29-7 |
| Assay Percent Range | >98% |
| Quantity | 50 mg |
| Molecular Formula | C10H11N3S2 |
| Formula Weight | Observed MW: 237.34 |