missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, 0.171 N Standardized Solution, Spectrum™ Chemical
Specifications
Specifications
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 20 L |
| Linear Formula | AgNO3 |
| Synonym | Nitric acid silver salt, Silver(I) nitrate |
| Solubility Information | Soluble in Water |
| PubChem CID | 24470 |
| Formula Weight | 169.87 g/mol |
| Concentration or Composition (by Analyte or Components) | 0.171 N ± 0.005 N |
| Odor | Odorless |
| Show More |
Safety and Handling
| Recommended Storage | Store in Light-resistant Containers |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.