missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ MPure™ Tissue DNA Extraction Kit
For extraction of genomic DNA from animal tissues, swab samples and blood stains
Supplier: MP Biomedicals™ 117022400
Description
MPure Tissue DNA Extraction Kit contains everything needed for sample preparation. However, for tough tissues such as cardiac or skeletal muscle, spleen, intestinal or other connective tissue-rich samples, sample preparation using FastPrep™ Instruments and Lysing Matrix Tubes can greatly increase yields as well as shorten processing time.
- Completely automated purification protocol
- Runs in as little as 40 minutes
- Produces nucleic acids that can be used immediately in downstream applications
Specifications
| Store at room temperature | |
| Magnetic Bead | |
| MPure-12 Automated Nucleic Acid Purification System | |
| Lysis Buffer 2, 720μL; Binding Buffer 1, 720μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer, 1000μL; Elution Buffer, 1000μL | |
| Purify DNA from 100 - 400μL or 10 - 40mg of tissue; 100 - 400μL or one swab or brush for dried swab samples; 100 - 400μL or four discs for dried blood |
| Tissue DNA Extraction Kit | |
| Animal Tissue, Blood Stain, Swab | |
| Tissue DNA Purification | |
| 48 Preps | |
| As little as 40 minutes |