missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ MPure™ Bacterial DNA Extraction Kit

Catalog No. 117022600
Change view
Click to view available options
No. of Reactions:
48 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
117022600 48 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 117022600 Supplier MP Biomedicals™ Supplier No. 117022600
Only null left

For extraction of genomic DNA from Gram-positive and Gram-negative bacteria

MPure Bacterial DNA Extraction Kit contains everything needed for sample preparation. However, for tough samples such as Gram-positive bacteria or spores, soil or feces, sample preparation using FastPrep™ Instruments and Lysing Matrix Tubes can increase yields as well as shorten processing time.

  • Starting materials include bacterial pellet/colony from culture, cell-free body fluids, liquid transport media, urine, and environmental material
  • Completely automated purification protocol
  • Run time as little as 55 minutes
  • DNA can be used immediately in downstream applications

Specifications

Content And Storage Store at room temperature
Description Bacterial DNA Extraction Kit
Isolation Technology Magnetic Bead
Sample Type Bacterial Cells
For Use With (Equipment) MPure-12 Automated Nucleic Acid Purification System
For Use With (Application) DNA Purification from Gram-positive and Gram-negative bacteria
Includes Proteinase K solution, 40μL; Lysis Buffer 2, 720μL; Binding Buffer 1, 720μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL
No. of Reactions 48 Preps
Sample Volume 200 - 400μL
Test Time As little as 55 minutes