missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mpro 13b-K
Description
Mpro 13b-K is a coronavirus Mpro inhibitor (IC50 values are 0.12, 0.58 and 0.9 μM for recombinant Mpro from SARS-CoV-2, MERS-CoV and SARS-CoV, respectively). Mpro 13b-K inhibits SARS-CoV-2 infection of human Calu3 cells (IC50 = 2.4 μM). The compound also inhibits SARS-CoV-2 replication in A549-ACE2-TMPRSS2, VeroE6 and Huh7 cell cultures (EC50 values are 0.84, 1.3 and 3.4 μM). Exhibits oral and inhalative bioavailability. Please note that this product was previously known as Mpro 13b.
Specifications
Specifications
| Chemical Name or Material | tert-Butyl (1-((S)-1-(((S)-4-(benzylamino)-3,4-dioxo-1-((S)-2-oxopyrrolidin-3-yl)butan-2-yl)amino)-3-cyclopropyl-1-oxopropan-2-yl)-2-oxo-1,2-dihydropyridin-3-yl)carbamate |
| CAS | 2412965-59-2 |
| Quantity | 5 mg |
| Product Line | Tocris |
| Target | 3C and 3CL Protease Inhibitors |
| Molecular Formula | C31H39N5O7 |
| Purity | 97% |