missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma mPAGE™ 8% Bis-Tris Precast Gel
Precast gel for research use only
$173.00 - $173.00
Specifications
| Buffer | MES or MOPS Running Buffer |
|---|---|
| Concentration | 8% Bis-Tris |
| For Use With (Equipment) | Mini Gel Systems |
| Gel Percentage | 8% |
| GelThickness | 1 mm |
| Catalog Number | Mfr. No. | Wells | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Wells | Price | Quantity | |||||
|
11-580-585
|
MilliporeSigma
MP8W10 |
10-well |
Pack of 10 Each for $173.00
|
|
|||||
|
11-580-606
|
MilliporeSigma
MP8W12 |
12-well |
Pack of 10 Each for $173.00
|
|
|||||
|
11-580-607
|
MilliporeSigma
MP8W15 |
15-well |
Pack of 10 Each for $173.00
|
|
|||||
Description
- mPAGE 8% Bis-Tris Precast Gel, 10x8 cm
- Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Specifications
| MES or MOPS Running Buffer | |
| Mini Gel Systems | |
| 1 mm | |
| Bis-Tris precast gels | |
| 10 Pk. | |
| 8 cm |
| 8% Bis-Tris | |
| 8% | |
| Bis-Tris | |
| Protein | |
| 10 cm |